Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIR2DL5A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174740.2 | NP_777165.1 | 3 - 4 | 357 | 91 | |
GenBank mRNA | AF490399.1 | - | 3 - 4 | 357 | 91 | |
AY075102.1 | - | 3 - 4 | 357 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Bos taurus Probe spans exons |