Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STATH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174752.3 | NP_777177.1 | 1 - 2 | 46 | 81 | |
GenBank mRNA | AY154893.1 | - | 1 - 2 | 46 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Bos taurus Probe spans exons |