Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175712.1 | NP_783643.1 | 4 - 5 | 2402 | 113 | |
GenBank mRNA | AF222713.2 | - | 4 - 5 | 2402 | 113 | |
BC120058.1 | - | 6 - 7 | 853 | 113 | ||
BT030564.1 | - | 4 - 5 | 623 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |