Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADGRE5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176661.1 | NP_788834.1 | 5 - 6 | 508 | 103 | |
GenBank mRNA | AJ416058.1 | - | 5 - 6 | 508 | 103 | |
AJ627191.1 | - | 7 - 8 | 839 | 103 | ||
BC149039.1 | - | 5 - 6 | 559 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |