Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: INHBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_176852.2 | NP_789822.2 | 1 - 2 | 454 | 89 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Bos taurus Probe spans exons |