Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FDPS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177497.2 | NP_803463.1 | 5 - 6 | 583 | 98 | |
GenBank mRNA | AF461050.1 | - | 5 - 6 | 583 | 98 | |
BC111256.1 | - | 6 - 7 | 98 | |||
BC120218.1 | - | 6 - 7 | 645 | 98 | ||
BC149572.1 | - | 6 - 7 | 704 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |