Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GGTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177511.2 | NP_803477.1 | 6 - 7 | 639 | 112 | |
GenBank mRNA | BC151691.1 | - | 5 - 6 | 351 | 112 | |
J04989.1 | - | 6 - 7 | 639 | 112 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |