Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ST3GAL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002892.2 | NP_001002892.1 | 5 - 6 | 1366 | 86 | |
GenBank mRNA | AJ748841.1 | - | 5 - 6 | 878 | 86 | |
BC105252.1 | - | 5 - 6 | 1366 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |