Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIDA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034208.2 | NP_001029380.1 | 2 - 3 | 250 | 91 | |
GenBank mRNA | BC102164.1 | - | 2 - 3 | 252 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |