Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANAPC15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034243.1 | NP_001029415.1 | 3 - 4 | 215 | 79 | |
GenBank mRNA | BC103197.1 | - | 3 - 4 | 215 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |