Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC28B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034305.2 | NP_001029477.1 | 1 - 2 | 59 | 95 | |
GenBank mRNA | BC102209.1 | - | 1 - 2 | 60 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Bos taurus Probe spans exons |