Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GATD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034463.2 | NP_001029635.1 | 2 - 3 | 230 | 85 | |
GenBank mRNA | BC102260.1 | - | 2 - 3 | 231 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |