Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034486.1 | NP_001029658.1 | 4 - 5 | 428 | 120 | |
GenBank mRNA | BC103312.1 | - | 4 - 5 | 428 | 120 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |