Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034493.1 | NP_001029665.1 | 3 - 4 | 303 | 135 | |
GenBank mRNA | BC102162.1 | - | 3 - 4 | 303 | 135 | |
BC134736.1 | - | 3 - 4 | 216 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |