Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDZD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034664.1 | NP_001029836.1 | 10 - 11 | 1499 | 83 | |
GenBank mRNA | BC105272.1 | - | 10 - 11 | 1499 | 83 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |