Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CALU | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034665.2 | NP_001029837.1 | 1 - 2 | 68 | 85 | |
GenBank mRNA | BC102361.1 | - | 1 - 2 | 69 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |