Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034674.1 | NP_001029846.1 | 2 - 3 | 126 | 75 | |
GenBank mRNA | BC102261.1 | - | 2 - 3 | 126 | 75 | |
BC148013.1 | - | 2 - 3 | 126 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |