Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRSF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034700.2 | NP_001029872.1 | 5 - 6 | 586 | 82 | |
GenBank mRNA | BC102735.1 | - | 5 - 6 | 588 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |