Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034714.2 | NP_001029886.1 | 5 - 6 | 1189 | 82 | |
GenBank mRNA | BC103343.1 | - | 5 - 6 | 1190 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |