Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034714.2 | NP_001029886.1 | 7 - 8 | 1394 | 107 | |
GenBank mRNA | BC103343.1 | - | 7 - 8 | 1395 | 107 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |