Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11H2orf42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034714.2 | NP_001029886.1 | 8 - 9 | 1559 | 93 | |
GenBank mRNA | BC103343.1 | - | 8 - 9 | 1560 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |