Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TAF12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034754.1 | NP_001029926.1 | 2 - 3 | 331 | 100 | |
GenBank mRNA | BC102082.1 | - | 2 - 3 | 331 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |