Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034765.1 | NP_001029937.1 | 1 - 2 | 48 | 66 | |
GenBank mRNA | BC104530.1 | - | 1 - 2 | 48 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Inventoried Probe spans exons |