Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034792.2 | NP_001029964.1 | 1 - 2 | 98 | 72 | |
GenBank mRNA | BC103340.1 | - | 1 - 2 | 99 | 72 | |
BT025461.1 | - | 1 - 2 | 106 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |