Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMA16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035034.2 | NP_001030206.1 | 4 - 5 | 273 | 131 | |
GenBank mRNA | BC102541.1 | - | 4 - 5 | 274 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |