Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPL22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035454.2 | NP_001030531.1 | 4 - 5 | 281 | 106 | |
GenBank mRNA | BC102665.1 | - | 4 - 5 | 282 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |