Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SMS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001035471.1 | NP_001030548.1 | 7 - 8 | 852 | 134 | |
GenBank mRNA | BC103012.1 | - | 7 - 8 | 852 | 134 | |
BC151416.1 | - | 7 - 8 | 778 | 134 | ||
BT029883.1 | - | 7 - 8 | 805 | 134 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |