Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040472.2 | NP_001035562.2 | 2 - 3 | 145 | 75 | |
GenBank mRNA | BC103010.1 | - | 2 - 3 | 141 | 75 | |
DV802141.1 | - | 2 - 3 | 134 | 75 | ||
EH165860.1 | - | 2 - 3 | 146 | 75 | ||
X53268.1 | - | 87 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |