Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040483.1 | NP_001035573.1 | 5 - 6 | 558 | 77 | |
GenBank mRNA | AF095935.1 | - | 2 - 3 | 229 | 77 | |
BC102067.1 | - | 5 - 6 | 558 | 77 | ||
DQ403015.1 | - | 4 - 5 | 469 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |