Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EEF1G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040487.2 | NP_001035577.1 | 9 - 10 | 1184 | 91 | |
GenBank mRNA | AB098752.1 | - | 1 - 2 | 238 | 91 | |
BC102691.1 | - | 9 - 10 | 1185 | 91 | ||
BT025451.1 | - | 8 - 9 | 1161 | 91 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |