Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC61G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040586.2 | NP_001035676.1 | 3 - 4 | 254 | 137 | |
GenBank mRNA | BC102186.1 | - | 3 - 4 | 255 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |