Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEX29 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076461.2 | NP_001069929.1 | 5 - 6 | 522 | 67 | |
GenBank mRNA | BC102719.1 | - | 5 - 6 | 523 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |