Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PHPT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076516.2 | NP_001069984.1 | 1 - 2 | 218 | 67 | |
GenBank mRNA | BC108196.1 | - | 1 - 2 | 220 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Bos taurus Inventoried Probe spans exons |