Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM166C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001076994.2 | NP_001070462.1 | 3 - 4 | 503 | 92 | |
GenBank mRNA | BC102738.1 | - | 3 - 4 | 504 | 92 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |