Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIIAD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077049.1 | NP_001070517.1 | 4 - 5 | 354 | 76 | |
GenBank mRNA | BC108187.1 | - | 4 - 5 | 354 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |