Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HSPA8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_174345.4 | NP_776770.2 | 2 - 3 | 281 | 69 | |
GenBank mRNA | BC105182.1 | - | 2 - 3 | 282 | 69 | |
BT030487.1 | - | 2 - 3 | 267 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |