Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: JCHAIN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175773.3 | NP_786967.1 | 2 - 3 | 221 | 83 | |
GenBank mRNA | BC103426.1 | - | 2 - 3 | 224 | 83 | |
L20311.1 | - | 2 - 3 | 194 | 83 | ||
U02301.1 | - | 2 - 3 | 194 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |