Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_203362.2 | NP_976238.2 | 1 - 2 | 337 | 114 | |
GenBank mRNA | BC102413.1 | - | 1 - 2 | 337 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |