Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DMAC2L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001007812.3 | NP_001007813.1 | 3 - 4 | 375 | 113 | |
GenBank mRNA | AY780292.1 | - | 3 - 4 | 364 | 113 | |
BC109757.1 | - | 3 - 4 | 377 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |