Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PSME1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001024509.3 | NP_001019680.2 | 9 - 10 | 700 | 63 | |
GenBank mRNA | BC105379.1 | - | 9 - 10 | 701 | 63 | |
DQ010410.1 | - | 9 - 10 | 618 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |