Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPL35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025321.3 | NP_001020492.1 | 74 | 76 | ||
GenBank mRNA | AY911347.1 | - | 2 - 3 | 76 | 76 | |
BC111654.1 | - | 75 | 76 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |