Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHMP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001191260.2 | NP_001178189.1 | 5 - 6 | 502 | 62 | |
GenBank mRNA | BC109956.1 | - | 5 - 6 | 503 | 62 | |
DN279295.1 | - | 3 - 4 | 672 | 62 | ||
DT837819.1 | - | 5 - 6 | 503 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |