Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037617.2 | NP_001032706.1 | 2 - 3 | 220 | 66 | |
GenBank mRNA | BC109792.1 | - | 2 - 3 | 221 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |