Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1H21orf91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037617.2 | NP_001032706.1 | 4 - 5 | 819 | 74 | |
GenBank mRNA | BC109792.1 | - | 4 - 5 | 820 | 74 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Bos taurus Probe spans exons |