Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ERP27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038041.2 | NP_001033130.1 | 1 - 2 | 177 | 66 | |
GenBank mRNA | BC109770.1 | - | 1 - 2 | 178 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Bos taurus Probe spans exons |