Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TEKT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038069.2 | NP_001033158.1 | 3 - 4 | 846 | 122 | |
GenBank mRNA | BC111132.1 | - | 3 - 4 | 847 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Bos taurus Probe spans exons |