Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFB2M | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038127.2 | NP_001033216.1 | 6 - 7 | 1014 | 75 | |
GenBank mRNA | BC109634.1 | - | 6 - 7 | 1015 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Bos taurus Probe spans exons |