Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SKA2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038579.2 | NP_001033668.1 | 1 - 2 | 54 | 69 | |
GenBank mRNA | BC109479.1 | - | 1 - 2 | 55 | 69 | |
BC146069.1 | - | 1 - 2 | 49 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Recommended Assay Bos taurus Probe spans exons |