Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C29H11orf98 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038581.2 | NP_001033670.1 | 3 - 4 | 306 | 72 | |
GenBank mRNA | BC109665.1 | - | 3 - 4 | 307 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Bos taurus Probe spans exons |