Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: THAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038669.1 | NP_001033758.1 | 4 - 5 | 1569 | 79 | |
GenBank mRNA | BC110168.1 | - | 4 - 5 | 1569 | 79 | |
BC118115.1 | - | 3 - 4 | 380 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Bos taurus Probe spans exons |